Search Thermo Fisher Scientific
Gene Symbol: AMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001633.3 | NP_001624.1 | 8 | 998 | ||
GenBank mRNA | AK290837.1 | - | 8 | 826 | ||
AK303795.1 | - | 8 | 724 | |||
AK310791.1 | - | 8 | 1015 | |||
AY544123.1 | - | 8 | 795 | |||
BC041593.1 | - | 8 | 961 | 5433460 | ||
X04225.1 | - | 8 | 807 | |||
X04494.1 | - | 8 | 780 |