Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142276.1 | NP_001135748.1 | 7 | 1159 | ||
NM_001243299.1 | NP_001230228.1 | 7 | 1077 | |||
NM_001642.2 | NP_001633.1 | 7 | 1159 | |||
GenBank mRNA | AF168956.1 | - | 7 | 1005 | ||
AK128162.1 | - | 7 | 1070 | |||
AK308248.1 | - | 5 | 703 | |||
BC000373.2 | - | 7 | 1059 | 2820109 | ||
L09209.1 | - | 7 | 1074 | |||
L23113.1 | - | 3 | 320 | |||
L27631.1 | - | 7 | 1075 | |||
S60099.1 | - | 7 | 1074 | |||
Z22572.1 | - | 7 | 1069 |