Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000047.2 | NP_000038.2 | 3 | 309 | ||
NM_001282628.1 | NP_001269557.1 | 4 | 595 | |||
XM_005274518.2 | XP_005274575.1 | 3 | 236 | |||
XM_005274519.4 | XP_005274576.1 | 3 | 399 | |||
XM_011545521.1 | XP_011543823.1 | 3 | 440 | |||
XM_017029525.1 | XP_016885014.1 | 4 | 526 | |||
XM_017029526.1 | XP_016885015.1 | 4 | 640 | |||
GenBank mRNA | AK223183.1 | - | 3 | 139 | ||
AK223199.1 | - | 3 | 106 | |||
AK301004.1 | - | 4 | 426 | |||
AK313093.1 | - | 3 | 128 | |||
BC130438.1 | - | 3 | 104 | 40146469 | ||
BP272004.1 | - | 3 | 241 | |||
BP272164.1 | - | 3 | 242 | |||
DA915010.1 | - | 3 | 311 | |||
DB073314.1 | - | 3 | 291 | |||
HX786884.1 | - | 3 | 200 | |||
X83573.1 | - | 3 | 126 |