Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DST SIRNA Assays ›
Gene Symbol:
DST
Gene Name:
dystonin
Gene Aliases:
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location:
Chr.6: 56457987 - 56954671 on Build GRCh38
UniGene ID:
Hs.604915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS101079
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144769.2
NP_001138241.1
28
3823
NM_001144770.1
NP_001138242.1
26
3494
NM_001723.5
NP_001714.1
15
2378
NM_015548.4
NP_056363.2
15
2378
NM_183380.3
NP_899236.1
25
3768
XM_005249310.3
XP_005249367.1
29
3917
XM_005249315.3
XP_005249372.1
28
3864
XM_005249316.2
XP_005249373.1
21
4140
XM_005249318.2
XP_005249375.1
27
3578
XM_005249319.2
XP_005249376.1
26
3479
XM_005249320.4
XP_005249377.1
26
3535
XM_005249322.4
XP_005249379.1
26
3430
XM_005249323.3
XP_005249380.1
25
3763
XM_005249324.2
XP_005249381.1
15
2375
XM_011514824.2
XP_011513126.1
28
3828
XM_011514825.2
XP_011513127.1
28
3824
XM_011514826.2
XP_011513128.1
25
3763
XM_017011205.1
XP_016866694.1
29
3927
XM_017011206.1
XP_016866695.1
29
3927
XM_017011207.1
XP_016866696.1
29
3864
XM_017011208.1
XP_016866697.1
29
3927
XM_017011209.1
XP_016866698.1
29
3927
XM_017011210.1
XP_016866699.1
29
3927
XM_017011211.1
XP_016866700.1
29
3927
XM_017011212.1
XP_016866701.1
29
3927
XM_017011213.1
XP_016866702.1
29
3927
XM_017011214.1
XP_016866703.1
29
3927
XM_017011215.1
XP_016866704.1
29
3927
XM_017011216.1
XP_016866705.1
29
3927
XM_017011217.1
XP_016866706.1
26
3508
XM_017011219.1
XP_016866708.1
29
3923
XM_017011220.1
XP_016866709.1
28
3866
XM_017011221.1
XP_016866710.1
29
3923
XM_017011222.1
XP_016866711.1
21
4082
XM_017011223.1
XP_016866712.1
29
3923
XM_017011224.1
XP_016866713.1
25
3763
XM_017011225.1
XP_016866714.1
29
3937
GenBank mRNA
AF400226.1
-
15
2270
AF400227.1
-
15
2270
AK299898.1
-
26
3494
AU117550.1
-
5
641
BC065536.1
-
15
2342
L11690.1
-
15
2372
M63618.1
-
5
609
M69225.1
-
15
2308
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
cell adhesion
integrin-mediated signaling pathway
response to wounding
maintenance of cell polarity
intermediate filament cytoskeleton organization
cell motility
actin binding
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0