Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFP36L2 SIRNA Assays ›
Gene Symbol:
ZFP36L2
Gene Name:
ZFP36 ring finger protein like 2
Gene Aliases:
BRF2, ERF-2, ERF2, RNF162C, TIS11D
Chromosome Location:
Chr.2: 43222402 - 43226606 on Build GRCh38
UniGene ID:
Hs.503093
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS101106
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFP36L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_006887.4
NP_008818.3
2
893
GenBank mRNA
BC005010.1
-
2
682
3640393
BC020369.1
-
2
882
X78992.1
-
2
668
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
regulation of transcription, DNA-templated
mRNA catabolic process
cell proliferation
response to wounding
hemopoiesis
T cell differentiation in thymus
somatic stem cell population maintenance
regulation of mRNA stability
cellular response to fibroblast growth factor stimulus
negative regulation of fat cell differentiation
somatic stem cell division
definitive hemopoiesis
3'-UTR-mediated mRNA destabilization
ERK1 and ERK2 cascade
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to glucocorticoid stimulus
cellular response to transforming growth factor beta stimulus
cellular response to granulocyte macrophage colony-stimulating factor stimulus
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
cellular response to phorbol 13-acetate 12-myristate
negative regulation of stem cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
mRNA 3'-UTR binding
protein binding
AU-rich element binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0