Search
Search

BTC| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316963.1 | NP_001303892.1 | 3 | 584 | ||
| NM_001729.3 | NP_001720.1 | 3 | 584 | |||
| XM_011532211.1 | XP_011530513.1 | 3 | 565 | |||
| GenBank mRNA | BC011618.2 | - | 3 | 375 | 3952444 | |
| KU177946.1 | - | 3 | 223 | |||
| KU177947.1 | - | 3 | 223 | |||
| S55606.1 | - | 3 | 517 | |||