Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS101440
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
6
867
NM_004912.3
NP_004903.2
7
1362
NM_194454.1
NP_919436.1
7
1015
NM_194455.1
NP_919437.1
7
1489
NM_194456.1
NP_919438.1
8
1254
XM_005250660.3
XP_005250717.1
7
982
XM_005250662.3
XP_005250719.1
7
961
XM_005250665.3
XP_005250722.1
7
889
XM_005250666.3
XP_005250723.1
8
1065
XM_005250667.2
XP_005250724.1
6
773
XM_005250668.3
XP_005250725.1
6
835
XM_005250669.3
XP_005250726.1
6
813
XM_006716161.3
XP_006716224.1
7
1113
XM_006716162.3
XP_006716225.1
7
930
XM_006716163.3
XP_006716226.1
6
780
XM_011516651.2
XP_011514953.1
8
1096
XM_011516653.2
XP_011514955.1
6
850
XM_011516654.2
XP_011514956.1
8
1114
XM_011516655.2
XP_011514957.1
6
768
XM_011516656.2
XP_011514958.1
6
737
XM_011516657.2
XP_011514959.1
6
657
XM_011516658.2
XP_011514960.1
6
669
XM_011516659.2
XP_011514961.1
6
691
XM_011516660.2
XP_011514962.1
5
573
XM_011516661.2
XP_011514963.1
5
651
XM_017012755.1
XP_016868244.1
6
740
XM_017012756.1
XP_016868245.1
6
702
XM_017012757.1
XP_016868246.1
6
734
XM_017012758.1
XP_016868247.1
7
1340
XM_017012759.1
XP_016868248.1
6
1192
GenBank mRNA
AF296765.2
-
7
1259
AF310133.1
-
7
954
AF388384.1
-
7
1489
AJ294850.1
-
8
1213
AK055305.1
-
6
867
AK056537.1
-
6
797
AY380057.1
-
7
954
AY380058.1
-
7
954
AY380059.1
-
7
954
AY380060.1
-
7
954
AY993944.1
-
4
470
BC094684.1
-
4
480
6174749
BX648659.1
-
3
335
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_030675, NM_001170552, AK039264, AF306509, BC085252, and others.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0