Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254750.1 | NP_001241679.1 | 5 | 1259 | ||
NM_001254751.1 | NP_001241680.1 | 5 | 1259 | |||
NM_006725.4 | NP_006716.3 | 5 | 1259 | |||
NR_045638.1 | 5 | 1259 | ||||
XM_006718738.1 | XP_006718801.1 | 5 | 1259 | |||
XM_006718739.1 | XP_006718802.1 | 5 | 1259 | |||
XM_006718740.1 | XP_006718803.1 | 5 | 1259 | |||
XM_006718741.1 | XP_006718804.1 | 5 | 1259 | |||
XM_011545360.1 | XP_011543662.1 | 5 | 1259 | |||
GenBank mRNA | AK128419.1 | - | 6 | 1163 | ||
AK292704.1 | - | 5 | 1135 | |||
AK312749.1 | - | 5 | 1220 | |||
BC033755.1 | - | 5 | 1175 | 5213263 | ||
BC078669.1 | - | 5 | 1198 | 6302947 | ||
U66142.1 | - | 5 | 1034 | |||
U66145.1 | - | 5 | 1034 | |||
U66146.1 | - | 5 | 1034 | |||
X60992.1 | - | 5 | 1154 |