siRNA Details
Gene Symbol: CHRNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000746.5 | NP_000737.1 | 2 | 190 | ||
NM_001190455.2 | NP_001177384.1 | 2 | 277 | |||
NR_046324.1 | 2 | 190 | ||||
XM_011521178.2 | XP_011519480.1 | 2 | 190 | |||
XM_017021882.1 | XP_016877371.1 | 2 | 128 | |||
XM_017021883.1 | XP_016877372.1 | 2 | 128 | |||
XM_017021884.1 | XP_016877373.1 | 2 | 211 | |||
GenBank mRNA | AF385585.1 | - | 2 | 92 | ||
AJ586911.1 | - | 2 | 95 | |||
AK292069.1 | - | 2 | 160 | |||
AK294229.1 | - | 2 | 272 | |||
AY909581.1 | - | 1 | 61 | |||
BC037571.1 | - | 2 | 117 | 5245812 | ||
DA583811.1 | - | 2 | 190 | |||
EF405802.1 | - | 2 | 128 | |||
EF405812.1 | - | 2 | 96 | |||
EF405813.1 | - | 2 | 96 | |||
EF405814.1 | - | 2 | 128 | |||
L25827.1 | - | 1 | 55 | |||
U40583.1 | - | 2 | 85 | |||
U62436.1 | - | 2 | 150 | |||
X70297.1 | - | 2 | 181 | |||
Y08420.1 | - | 2 | 78 |

More Information
Panther Classification:
Molecular Function -






