siRNA Details
Gene Symbol: CHRNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000746.5 | NP_000737.1 | 4 | 437 | ||
NM_001190455.2 | NP_001177384.1 | 4 | 524 | |||
XM_011521176.2 | XP_011519478.2 | 3 | 292 | |||
XM_011521178.2 | XP_011519480.1 | 4 | 437 | |||
XM_017021882.1 | XP_016877371.1 | 4 | 375 | |||
XM_017021883.1 | XP_016877372.1 | 4 | 375 | |||
GenBank mRNA | AF385585.1 | - | 4 | 339 | ||
AJ586911.1 | - | 4 | 342 | |||
AK292069.1 | - | 4 | 407 | |||
AK294229.1 | - | 4 | 519 | |||
AY909581.1 | - | 3 | 307 | |||
DA583811.1 | - | 4 | 437 | |||
EF405802.1 | - | 4 | 375 | |||
EF405804.1 | - | 3 | 184 | |||
EF405805.1 | - | 1 | 49 | |||
EF405812.1 | - | 4 | 343 | |||
EF405813.1 | - | 4 | 333 | |||
EF405814.1 | - | 4 | 375 | |||
EF405815.1 | - | 3 | 154 | |||
EF405819.1 | - | 3 | 182 | |||
L25827.1 | - | 3 | 302 | |||
U40583.1 | - | 4 | 332 | |||
U62436.1 | - | 4 | 397 | |||
X70297.1 | - | 4 | 428 | |||
Y08420.1 | - | 4 | 325 | |||
Z23141.1 | - | 3 | 256 |

More Information
Panther Classification:
Molecular Function -






