Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CNR1 SIRNA Assays ›
          Gene Symbol:
          CNR1
          Gene Name:
          cannabinoid receptor 1
          Gene Aliases:
          CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNR
          Chromosome Location:
          Chr.6: 88139864 - 88167429 on Build GRCh38
          UniGene ID:
          Hs.75110
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102082
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CNR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001160226.1 NP_001153698.1 3 1532
          NM_001160258.1 NP_001153730.1 4 1570
          NM_001160259.1 NP_001153731.1 2 1445
          NM_016083.4 NP_057167.2 2 1389
          NM_033181.3 NP_149421.2 2 1044
          XM_006715330.3 XP_006715393.1 2 1336
          XM_011535424.1 XP_011533726.1 3 1876
          XM_011535425.1 XP_011533727.1 4 1914
          XM_011535426.1 XP_011533728.1 5 2072
          XM_011535428.1 XP_011533730.1 3 1723
          XM_017010238.1 XP_016865727.1 4 4700
          XM_017010239.1 XP_016865728.1 3 3023
          XM_017010240.1 XP_016865729.1 2 1383
          GenBank mRNA AK313908.1 - 2 1210
          AY766182.1 - 2 981
          AY995204.1 - 1 1080
          BC074811.2 - 1 1118 30915316
          BC074812.2 - 1 1118 30915532
          BC095513.1 - 1 1077
          BC100968.1 - 1 1080 40004970
          BC100969.1 - 1 1080 40004975
          BC100970.1 - 1 1080 40004978
          BC100971.1 - 1 1080 40004979
          DQ067455.1 - 1 1080
          KC292264.1 - 2 1044
          X54937.1 - 2 1228
          X81120.1 - 1 1171
          X81121.1 - 2 913

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of acute inflammatory response to antigenic stimulus
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          spermatogenesis
          axonal fasciculation
          aging
          response to nutrient
          memory
          positive regulation of neuron projection development
          sensory perception of pain
          positive regulation of fever generation
          negative regulation of fatty acid beta-oxidation
          regulation of synaptic transmission, GABAergic
          response to lipopolysaccharide
          negative regulation of mast cell activation
          negative regulation of dopamine secretion
          response to nicotine
          cannabinoid signaling pathway
          response to cocaine
          glucose homeostasis
          positive regulation of apoptotic process
          negative regulation of ion transport
          response to morphine
          response to ethanol
          negative regulation of action potential
          negative regulation of blood pressure
          positive regulation of blood pressure
          regulation of insulin secretion
          negative regulation of nitric-oxide synthase activity
          regulation of synaptic transmission, glutamatergic
          maternal process involved in female pregnancy
          regulation of feeding behavior
          regulation of penile erection
          cannabinoid receptor activity
          drug binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tt4lt:80/100.66.77.34:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0