Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031847.2 | NP_001027017.1 | 11 | 1423 | ||
NM_001876.3 | NP_001867.2 | 11 | 1423 | |||
GenBank mRNA | AK025307.1 | - | 6 | 709 | ||
AK172798.1 | - | 11 | 1409 | |||
AK309464.1 | - | 11 | 1399 | |||
AK314301.1 | - | 11 | 1362 | |||
BC000185.1 | - | 11 | 1298 | 3352642 | ||
BT009791.1 | - | 10 | 1253 | |||
L39211.1 | - | 11 | 1329 |