Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CREB1 SIRNA Assays ›
          Gene Symbol:
          CREB1
          Gene Name:
          cAMP responsive element binding protein 1
          Gene Aliases:
          CREB, CREB-1
          Chromosome Location:
          Chr.2: 207529892 - 207605989 on Build GRCh38
          UniGene ID:
          Hs.516646
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102262
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CREB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320793.1 NP_001307722.1 3 397
          NM_004379.4 NP_004370.1 3 397
          NM_134442.4 NP_604391.1 3 397
          NR_135473.1 3 397
          XM_011510645.1 XP_011508947.1 3 211
          XM_011510646.2 XP_011508948.1 3 373
          XM_011510647.2 XP_011508949.1 3 254
          XM_011510648.2 XP_011508950.1 3 373
          XM_011510649.2 XP_011508951.1 2 208
          XM_011510650.2 XP_011508952.1 2 307
          XM_011510651.2 XP_011508953.1 2 166
          XM_017003399.1 XP_016858888.1 3 347
          XM_017003400.1 XP_016858889.1 2 253
          GenBank mRNA AK298740.1 - 3 219
          AK302333.1 - 2 166
          AK312747.1 - 4 292
          AK315226.1 - 3 188
          AY347527.1 - 3 189
          BC010636.1 - 3 240 3872792
          BC095407.1 - 3 341 30344522
          BT019509.1 - 2 146
          CR407617.1 - 2 146
          DC366797.1 - 3 397
          M27691.1 - 3 262
          M34356.1 - 2 146
          X55545.1 - 289
          X60003.1 - 2 146

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          transcription from RNA polymerase II promoter
          protein phosphorylation
          signal transduction
          transforming growth factor beta receptor signaling pathway
          axon guidance
          aging
          lactation
          memory
          circadian rhythm
          regulation of cell size
          visual learning
          response to organic substance
          negative regulation of transcription by competitive promoter binding
          response to activity
          viral process
          pituitary gland development
          positive regulation of transforming growth factor beta3 production
          secretory granule organization
          response to glucagon
          chemotaxis to arachidonic acid
          response to nicotine
          cellular response to hepatocyte growth factor stimulus
          cellular response to platelet-derived growth factor stimulus
          response to antidepressant
          positive regulation of multicellular organism growth
          regulation of circadian rhythm
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          positive regulation of osteoclast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hormone secretion
          positive regulation of lipid biosynthetic process
          regulation of fibroblast proliferation
          protein stabilization
          regulation of glial cell proliferation
          lung saccule development
          Type I pneumocyte differentiation
          cellular response to zinc ion
          cellular response to fatty acid
          positive regulation of long-term synaptic potentiation
          negative regulation of neuron death
          response to L-glutamate
          cellular response to nerve growth factor stimulus
          cellular response to insulin-like growth factor stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription cofactor activity
          protein binding
          enzyme binding
          Hsp70 protein binding
          histone acetyltransferase binding
          cAMP response element binding
          identical protein binding
          arrestin family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0