Search
Search

CRYGD| Gene Symbol: CRYGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006891.3 | NP_008822.2 | 3 | 620 | ||
| GenBank mRNA | BC117338.1 | - | 3 | 620 | 40125889 | |
| BC117340.1 | - | 3 | 620 | 40125891 | ||
| CD674533.1 | - | 2 | 484 | |||
| EL946552.1 | - | 3 | 536 | |||
| KT966392.1 | - | 3 | 528 | |||
| U66583.1 | - | 2 | 504 | |||