Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017541.2 | NP_060011.1 | 2, 3 | 327 | ||
GenBank mRNA | AF161703.1 | - | 2, 3 | 319 | ||
AK314172.1 | - | 2, 3 | 327 | |||
AL133079.1 | - | 1, 2 | 1177 | |||
BC069478.1 | - | 2, 3 | 325 | 7262271 | ||
BC070241.1 | - | 3, 4 | 615 | 30405628 | ||
KT966393.1 | - | 2, 3 | 315 | |||
L36869.1 | - | 1, 2 | 218 |