Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCTD SIRNA Assays ›
Gene Symbol:
DCTD
Gene Name:
dCMP deaminase
Gene Aliases:
-
Chromosome Location:
Chr.4: 182890091 - 182917477 on Build GRCh38
UniGene ID:
Hs.183850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS102678
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCTD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012732.1
NP_001012750.1
5, 6
656
NM_001921.2
NP_001912.2
5, 6
633
XM_005262778.3
XP_005262835.1
5, 6
681
XM_005262779.3
XP_005262836.1
6, 7
772
XM_005262780.3
XP_005262837.1
7, 8
780
XM_005262781.4
XP_005262838.1
4, 5
438
XM_005262782.3
XP_005262839.1
4, 5
435
XM_006714115.3
XP_006714178.1
5, 6
579
XM_006714116.3
XP_006714179.1
4, 5
876
XM_011531674.2
XP_011529976.1
6, 7
1266
XM_011531675.2
XP_011529977.1
5, 6
997
XM_011531676.2
XP_011529978.1
6, 7
1005
XM_017007816.1
XP_016863305.1
6, 7
748
XM_017007817.1
XP_016863306.1
6, 7
715
XM_017007818.1
XP_016863307.1
7, 8
780
XM_017007819.1
XP_016863308.1
7, 8
1274
XM_017007820.1
XP_016863309.1
7, 8
1137
XM_017007821.1
XP_016863310.1
7, 8
1270
XM_017007822.1
XP_016863311.1
7, 8
615
XM_017007823.1
XP_016863312.1
4, 5
726
XM_017007824.1
XP_016863313.1
4, 5
353
GenBank mRNA
AK293135.1
-
4, 5
412
AK310788.1
-
4, 5
416
AK313221.1
-
5, 6
558
BC001286.1
-
5, 6
508
3458999
BC088357.1
-
5, 6
547
30389169
BQ228061.1
-
5, 6
751
BU188090.1
-
4, 5
462
L12136.1
-
4, 5
459
Back To Top
More Information
Panther Classification:
Molecular Function -
deaminase
hydrolase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleotide metabolic process
nucleotide biosynthetic process
pyrimidine nucleoside biosynthetic process
dCMP deaminase activity
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0