Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290321.2 | NP_001277250.1 | 17 | 3223 | ||
NM_001290322.2 | NP_001277251.1 | 17 | 4531 | |||
NM_005509.5 | NP_005500.4 | 17 | 3223 | |||
XM_005271909.4 | XP_005271966.1 | 18 | 2921 | |||
XM_005271910.4 | XP_005271967.1 | 17 | 3266 | |||
XM_005271912.2 | XP_005271969.1 | 18 | 2905 | |||
XM_011543212.2 | XP_011541514.1 | 17 | 3266 | |||
XM_011543213.2 | XP_011541515.1 | 18 | 2851 | |||
XM_011543214.2 | XP_011541516.1 | 14 | 2432 | |||
XM_011543215.2 | XP_011541517.1 | 13 | 2268 | |||
XM_017009141.1 | XP_016864630.1 | 18 | 2921 | |||
XM_017009142.1 | XP_016864631.1 | 18 | 2851 | |||
XM_017009143.1 | XP_016864632.1 | 19 | 3100 | |||
XM_017009144.1 | XP_016864633.1 | 17 | 2803 | |||
XM_017009145.1 | XP_016864634.1 | 17 | 2673 | |||
XM_017009146.1 | XP_016864635.1 | 17 | 4115 | |||
XM_017009147.1 | XP_016864636.1 | 17 | 3266 | |||
XM_017009148.1 | XP_016864637.1 | 16 | 3069 | |||
GenBank mRNA | AB593142.1 | - | 17 | 4534 | ||
AJ005821.1 | - | 17 | 2820 | |||
BC144537.1 | - | 17 | 2766 | 9053064 | ||
BC150616.1 | - | 17 | 2766 | 9056986 |