Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079911.2 | NP_001073380.1 | 5 | 691 | ||
NM_004407.3 | NP_004398.1 | 6 | 739 | |||
XM_011531705.2 | XP_011530007.1 | 6 | 1614 | |||
XM_011531706.2 | XP_011530008.1 | 5 | 1566 | |||
GenBank mRNA | BC130581.1 | - | 5 | 661 | 40146612 | |
BC132865.1 | - | 5 | 661 | 40146887 | ||
U65378.1 | - | 5 | 686 | |||
U89012.1 | - | 6 | 734 |