Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS103057
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
4
1162
NM_057159.2
NP_476500.1
5
1240
XM_005251781.3
XP_005251838.1
5
1385
XM_005251782.3
XP_005251839.1
5
1479
XM_017014383.1
XP_016869872.1
5
1279
XM_017014384.1
XP_016869873.1
6
1653
XM_017014385.1
XP_016869874.1
7
1246
XM_017014386.1
XP_016869875.1
7
1546
XM_017014387.1
XP_016869876.1
7
1282
XM_017014388.1
XP_016869877.1
7
1465
XM_017014389.1
XP_016869878.1
6
1208
XM_017014390.1
XP_016869879.1
7
1243
XM_017014391.1
XP_016869880.1
7
1267
XM_017014392.1
XP_016869881.1
6
1510
XM_017014393.1
XP_016869882.1
7
1306
XM_017014394.1
XP_016869883.1
6
1435
XM_017014395.1
XP_016869884.1
6
1263
XM_017014396.1
XP_016869885.1
5
1205
XM_017014397.1
XP_016869886.1
6
1136
XM_017014398.1
XP_016869887.1
6
1379
XM_017014399.1
XP_016869888.1
6
1134
XM_017014400.1
XP_016869889.1
7
1495
XM_017014401.1
XP_016869890.1
6
1166
XM_017014402.1
XP_016869891.1
6
1223
XM_017014403.1
XP_016869892.1
7
1221
XM_017014404.1
XP_016869893.1
5
1419
XM_017014405.1
XP_016869894.1
7
1408
XM_017014406.1
XP_016869895.1
6
1142
XM_017014407.1
XP_016869896.1
6
1198
GenBank mRNA
AK022808.1
-
4
1162
AK290287.1
-
6
1515
AK296374.1
-
3
843
AK298590.1
-
3
819
AK299279.1
-
3
893
AY322546.1
-
3
814
BC030615.2
-
5
1069
4815446
BC036034.1
-
5
1199
5272431
BC073167.1
-
6
1109
6193969
U78192.1
-
4
936
U80811.1
-
3
840
Y09479.1
-
4
936
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5rpk4:80/100.66.77.5:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0