Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELAVL2 SIRNA Assays ›
          Gene Symbol:
          ELAVL2
          Gene Name:
          ELAV like neuron-specific RNA binding protein 2
          Gene Aliases:
          HEL-N1, HELN1, HUB
          Chromosome Location:
          Chr.9: 23690099 - 23850757 on Build GRCh38
          UniGene ID:
          Hs.166109
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103171
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ELAVL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171195.1 NP_001164666.1 5 765
          NM_001171197.1 NP_001164668.1 5 778
          NM_004432.3 NP_004423.2 5 784
          XM_005251393.3 XP_005251450.1 5 922
          XM_005251394.3 XP_005251451.1 5 1079
          XM_005251395.3 XP_005251452.1 5 859
          XM_006716734.3 XP_006716797.1 6 1171
          XM_006716735.3 XP_006716798.1 6 639
          XM_006716736.3 XP_006716799.1 5 1060
          XM_011517774.2 XP_011516076.1 6 1185
          XM_011517776.2 XP_011516078.1 5 615
          XM_011517777.2 XP_011516079.1 6 1171
          XM_011517778.2 XP_011516080.2 5 708
          XM_011517779.2 XP_011516081.1 6 683
          XM_011517780.2 XP_011516082.1 6 1012
          XM_011517783.2 XP_011516085.1 6 1045
          XM_011517784.2 XP_011516086.1 5 740
          XM_011517785.2 XP_011516087.1 5 545
          XM_011517786.2 XP_011516088.1 5 919
          XM_017014408.1 XP_016869897.1 7 1186
          XM_017014409.1 XP_016869898.1 6 824
          XM_017014410.1 XP_016869899.1 6 968
          XM_017014411.1 XP_016869900.1 6 1151
          XM_017014412.1 XP_016869901.1 7 1133
          XM_017014413.1 XP_016869902.1 7 1132
          XM_017014414.1 XP_016869903.1 6 1155
          XM_017014415.1 XP_016869904.1 5 601
          XM_017014416.1 XP_016869905.1 6 674
          XM_017014417.1 XP_016869906.1 6 1045
          XM_017014418.1 XP_016869907.1 5 879
          XM_017014419.1 XP_016869908.1 5 770
          XM_017014420.1 XP_016869909.1 5 773
          XM_017014421.1 XP_016869910.1 5 891
          XM_017014422.1 XP_016869911.1 5 1060
          XM_017014423.1 XP_016869912.1 5 717
          XM_017014424.1 XP_016869913.1 5 1063
          XM_017014425.1 XP_016869914.1 5 871
          XM_017014426.1 XP_016869915.1 4 400
          GenBank mRNA AB209294.1 - 5 784
          BC030692.1 - 5 782 4826082
          BC042393.1 - 5 768

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_006238385, XM_006238388, XM_008763850, XM_008763848, XM_006238389, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001177883, NM_207686, NM_207685, NM_010486, U29088, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          regulation of transcription, DNA-templated
          nucleotide binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9j9pp:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0