Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193503.1 | NP_001180432.1 | 6 | 802 | ||
NM_001976.4 | NP_001967.3 | 8 | 973 | |||
NM_053013.3 | NP_443739.3 | 8 | 931 | |||
XM_005256521.2 | XP_005256578.1 | 8 | 1359 | |||
XM_011523729.1 | XP_011522031.1 | 8 | 949 | |||
XM_017024346.1 | XP_016879835.1 | 8 | 1366 | |||
GenBank mRNA | AK300662.1 | - | 8 | 817 | ||
AK300673.1 | - | 7 | 793 | |||
AK300709.1 | - | 6 | 772 | |||
AK309864.1 | - | 9 | 1024 | |||
BC017249.2 | - | 8 | 888 | 4902149 | ||
X16504.1 | - | 7 | 835 | |||
X51957.1 | - | 7 | 847 |