Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
siRNA ID
HSS103264
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
6
1014
NM_001166006.1
NP_001159478.1
6
1087
NM_001166007.1
NP_001159479.1
6
997
NM_004437.3
NP_004428.1
7
1077
NM_203342.2
NP_976217.1
7
1094
NM_203343.2
NP_976218.1
5
909
XM_005245753.1
XP_005245810.1
6
1089
XM_005245757.1
XP_005245814.1
6
1089
XM_005245760.1
XP_005245817.1
6
1089
XM_005245761.1
XP_005245818.1
6
1089
XM_005245763.1
XP_005245820.1
6
1089
XM_005245764.1
XP_005245821.1
6
1089
XM_005245765.1
XP_005245822.1
6
1089
XM_005245768.1
XP_005245825.1
6
1089
XM_005245769.1
XP_005245826.1
6
1089
XM_005245770.1
XP_005245827.1
6
1089
XM_005245772.4
XP_005245829.1
6
1089
XM_005245773.4
XP_005245830.1
6
1089
XM_005245774.1
XP_005245831.1
6
1089
XM_006710434.1
XP_006710497.1
6
1089
XM_006710439.1
XP_006710502.1
6
1089
XM_011540956.1
XP_011539258.1
6
1089
XM_011540957.1
XP_011539259.1
6
1089
XM_011540958.1
XP_011539260.1
6
1089
XM_011540959.1
XP_011539261.1
6
1089
XM_011540960.1
XP_011539262.1
6
1089
XM_011540961.1
XP_011539263.1
6
1089
XM_011540962.1
XP_011539264.1
6
1089
XM_011540963.2
XP_011539265.1
6
1089
XM_011540964.1
XP_011539266.1
6
985
XM_011540965.2
XP_011539267.1
6
1089
XM_017000581.1
XP_016856070.1
5
984
XM_017000582.1
XP_016856071.1
5
984
XM_017000583.1
XP_016856072.1
6
1089
XM_017000584.1
XP_016856073.1
6
1380
XM_017000585.1
XP_016856074.1
6
1377
XM_017000586.1
XP_016856075.1
6
1089
XM_017000587.1
XP_016856076.1
6
1089
XM_017000588.1
XP_016856077.1
6
1089
XM_017000589.1
XP_016856078.1
5
984
XM_017000590.1
XP_016856079.1
5
984
XM_017000591.1
XP_016856080.1
6
1089
XM_017000592.1
XP_016856081.1
6
1089
XM_017000593.1
XP_016856082.1
6
985
XM_017000594.1
XP_016856083.1
6
985
XM_017000595.1
XP_016856084.1
6
985
XM_017000596.1
XP_016856085.1
6
985
XM_017000597.1
XP_016856086.1
6
985
XM_017000598.1
XP_016856087.1
6
985
XM_017000599.1
XP_016856088.1
6
985
XM_017000600.1
XP_016856089.1
6
985
XM_017000601.1
XP_016856090.1
6
985
XM_017000602.1
XP_016856091.1
6
985
XM_017000603.1
XP_016856092.1
6
985
XM_017000604.1
XP_016856093.1
6
985
GenBank mRNA
AB209649.1
-
6
997
AF156225.1
-
5
887
AK310655.1
-
5
925
BC039079.1
-
6
1084
5167140
BC096104.1
-
3
303
BC096105.1
-
5
621
40001729
BC113841.1
-
5
621
BC114450.1
-
5
621
J03796.1
-
5
828
M14993.1
-
7
1058
M61733.1
-
7
1075
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0