Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
siRNA ID
HSS103266
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
5, 6
957
NM_001166006.1
NP_001159478.1
5, 6
1030
NM_001166007.1
NP_001159479.1
5, 6
940
NM_004437.3
NP_004428.1
6, 7
1020
NM_203342.2
NP_976217.1
6, 7
1037
NM_203343.2
NP_976218.1
4, 5
852
XM_005245753.1
XP_005245810.1
5, 6
1032
XM_005245757.1
XP_005245814.1
5, 6
1032
XM_005245760.1
XP_005245817.1
5, 6
1032
XM_005245761.1
XP_005245818.1
5, 6
1032
XM_005245763.1
XP_005245820.1
5, 6
1032
XM_005245764.1
XP_005245821.1
5, 6
1032
XM_005245765.1
XP_005245822.1
5, 6
1032
XM_005245768.1
XP_005245825.1
5, 6
1032
XM_005245769.1
XP_005245826.1
5, 6
1032
XM_005245770.1
XP_005245827.1
5, 6
1032
XM_005245772.4
XP_005245829.1
5, 6
1032
XM_005245773.4
XP_005245830.1
5, 6
1032
XM_005245774.1
XP_005245831.1
5, 6
1032
XM_006710434.1
XP_006710497.1
5, 6
1032
XM_006710439.1
XP_006710502.1
5, 6
1032
XM_011540956.1
XP_011539258.1
5, 6
1032
XM_011540957.1
XP_011539259.1
5, 6
1032
XM_011540958.1
XP_011539260.1
5, 6
1032
XM_011540959.1
XP_011539261.1
5, 6
1032
XM_011540960.1
XP_011539262.1
5, 6
1032
XM_011540961.1
XP_011539263.1
5, 6
1032
XM_011540962.1
XP_011539264.1
5, 6
1032
XM_011540963.2
XP_011539265.1
5, 6
1032
XM_011540964.1
XP_011539266.1
5, 6
928
XM_011540965.2
XP_011539267.1
5, 6
1032
XM_017000581.1
XP_016856070.1
4, 5
927
XM_017000582.1
XP_016856071.1
4, 5
927
XM_017000583.1
XP_016856072.1
5, 6
1032
XM_017000584.1
XP_016856073.1
5, 6
1323
XM_017000585.1
XP_016856074.1
5, 6
1320
XM_017000586.1
XP_016856075.1
5, 6
1032
XM_017000587.1
XP_016856076.1
5, 6
1032
XM_017000588.1
XP_016856077.1
5, 6
1032
XM_017000589.1
XP_016856078.1
4, 5
927
XM_017000590.1
XP_016856079.1
4, 5
927
XM_017000591.1
XP_016856080.1
5, 6
1032
XM_017000592.1
XP_016856081.1
5, 6
1032
XM_017000593.1
XP_016856082.1
5, 6
928
XM_017000594.1
XP_016856083.1
5, 6
928
XM_017000595.1
XP_016856084.1
5, 6
928
XM_017000596.1
XP_016856085.1
5, 6
928
XM_017000597.1
XP_016856086.1
5, 6
928
XM_017000598.1
XP_016856087.1
5, 6
928
XM_017000599.1
XP_016856088.1
5, 6
928
XM_017000600.1
XP_016856089.1
5, 6
928
XM_017000601.1
XP_016856090.1
5, 6
928
XM_017000602.1
XP_016856091.1
5, 6
928
XM_017000603.1
XP_016856092.1
5, 6
928
XM_017000604.1
XP_016856093.1
5, 6
928
GenBank mRNA
AB209649.1
-
5, 6
940
AF156225.1
-
4, 5
830
AK310655.1
-
4, 5
868
BC039079.1
-
5, 6
1027
5167140
BC096104.1
-
2, 3
246
BC096105.1
-
4, 5
564
40001729
BC113841.1
-
4, 5
564
BC114450.1
-
4, 5
564
J03796.1
-
4, 5
771
M14993.1
-
6, 7
1001
M61733.1
-
6, 7
1018
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0