Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004457.3 | NP_004448.2 | 8, 9 | 1416 | ||
NM_203372.1 | NP_976251.1 | 7, 8 | 1309 | |||
XM_017003584.1 | XP_016859073.1 | 6, 7 | 1228 | |||
GenBank mRNA | AB061712.1 | - | 8, 9 | 1384 | ||
AK001471.1 | - | 3, 4 | 335 | |||
AK023191.1 | - | 5, 6 | 961 | |||
BC041692.1 | - | 8, 9 | 1408 | 5226905 | ||
D89053.1 | - | 6, 7 | 1107 |