Search
Search

G6PD| Gene Symbol: G6PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000402.4 | NP_000393.4 | 10 | 1464 | ||
| NM_001042351.2 | NP_001035810.1 | 10 | 1353 | |||
| GenBank mRNA | AK292304.1 | - | 10 | 1290 | ||
| AK302341.1 | - | 10 | 1291 | |||
| BC000337.2 | - | 10 | 1279 | 2822640 | ||
| M21248.1 | - | 9 | 1240 | |||
| X03674.1 | - | 10 | 1696 | |||