Search
Search

GABRA3| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000808.3 | NP_000799.1 | 3 | 464 | ||
| XM_006724811.2 | XP_006724874.1 | 3 | 464 | |||
| GenBank mRNA | A28102.1 | - | 3 | 310 | ||
| AK312404.1 | - | 3 | 442 | |||
| BC028629.1 | - | 3 | 463 | 5261326 | ||
| DA402645.1 | - | 3 | 464 | |||
| KJ620007.1 | - | 9 | 1093 | |||
| S62908.1 | - | 3 | 309 | |||