siRNA Details
Gene Symbol: GABRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291985.1 | NP_001278914.1 | 9 | 1252 | ||
NM_014211.2 | NP_055026.1 | 10 | 1440 | |||
XM_005265872.2 | XP_005265929.1 | 8 | 1159 | |||
GenBank mRNA | AK055205.1 | - | 10 | 1079 | ||
AK292901.1 | - | 10 | 1434 | |||
AK296805.1 | - | 6 | 907 | |||
AK300303.1 | - | 9 | 1246 | |||
BC074810.2 | - | 9 | 1277 | 30915531 | ||
BC074865.2 | - | 9 | 1277 | 30915230 | ||
BC109105.1 | - | 10 | 1365 | 40035263 | ||
BC109106.1 | - | 10 | 1365 | 40035265 | ||
U95367.1 | - | 10 | 1398 |

More Information
Panther Classification:
Molecular Function -






