Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPC3 SIRNA Assays ›
          Gene Symbol:
          GPC3
          Gene Name:
          glypican 3
          Gene Aliases:
          DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
          Chromosome Location:
          Chr.X: 133535745 - 133985646 on Build GRCh38
          UniGene ID:
          Hs.644108
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS104149
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GPC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164617.1 NP_001158089.1 8 1795
          NM_001164618.1 NP_001158090.1 7 1678
          NM_001164619.1 NP_001158091.1 6 1564
          NM_004484.3 NP_004475.1 7 1726
          GenBank mRNA AK222761.1 - 7 1674
          AK222766.1 - 7 1674
          AK300168.1 - 7 1649
          AK310196.1 - 6 1393
          BC035972.1 - 7 1674 4603748
          BX329946.2 - 2 606
          DQ349136.1 - 6 1367
          DQ349137.1 - 7 1529
          DQ349138.1 - 8 1598
          JQ943686.1 - 7 1529
          L47125.1 - 7 1680
          L47176.1 - 7 1675
          Z37987.1 - 7 1637

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          branching involved in ureteric bud morphogenesis
          glycosaminoglycan biosynthetic process
          glycosaminoglycan catabolic process
          anatomical structure morphogenesis
          anterior/posterior axis specification
          body morphogenesis
          negative regulation of peptidase activity
          glycosaminoglycan metabolic process
          bone mineralization
          osteoclast differentiation
          lung development
          positive regulation of BMP signaling pathway
          embryonic hindlimb morphogenesis
          positive regulation of protein catabolic process
          positive regulation of endocytosis
          negative regulation of smoothened signaling pathway
          positive regulation of smoothened signaling pathway
          negative regulation of growth
          positive regulation of glucose import
          negative regulation of epithelial cell proliferation
          coronary vasculature development
          mesenchymal cell proliferation involved in ureteric bud development
          mesonephric duct morphogenesis
          cell proliferation involved in metanephros development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          protein binding
          heparan sulfate proteoglycan binding
          peptidyl-dipeptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0