Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 3 | 456 | ||
NM_001167607.1 | NP_001161079.1 | 3 | 456 | |||
GenBank mRNA | AK225978.1 | - | 3 | 384 | ||
AK298867.1 | - | 3 | 323 | |||
AK312494.1 | - | 3 | 351 | |||
BC009273.2 | - | 3 | 320 | 4053681 | ||
BX473154.1 | - | 3 | 434 | |||
L01439.1 | - | 3 | 381 | |||
M76477.1 | - | 3 | 381 | |||
X16087.1 | - | 3 | 249 | |||
X61094.1 | - | 2 | 166 | |||
X61095.1 | - | 3 | 312 | |||
X62078.1 | - | 3 | 349 |