Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GNAS SIRNA Assays ›
Gene Symbol:
GNAS
Gene Name:
GNAS complex locus
Gene Aliases:
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
Chromosome Location:
Chr.20: 58839681 - 58911196 on Build GRCh38
UniGene ID:
Hs.125898
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS104240
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GNAS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000516.5
NP_000507.1
8
1034
NM_001077488.3
NP_001070956.1
9
1037
NM_001077489.3
NP_001070957.1
7
989
NM_001077490.2
NP_001070958.1
8
2824
NM_001309840.1
NP_001296769.1
8
700
NM_001309861.1
NP_001296790.1
8
745
NM_016592.3
NP_057676.1
9
1643
NM_080425.3
NP_536350.2
8
2824
NM_080426.3
NP_536351.1
8
992
XM_017027812.1
XP_016883301.1
9
3094
XM_017027813.1
XP_016883302.1
8
3049
XM_017027814.1
XP_016883303.1
7
3046
XM_017027815.1
XP_016883304.1
7
517
XM_017027816.1
XP_016883305.1
9
1322
XM_017027817.1
XP_016883306.1
7
604
XM_017027818.1
XP_016883307.1
7
527
XM_017027819.1
XP_016883308.1
7
568
XM_017027820.1
XP_016883309.1
7
508
XM_017027821.1
XP_016883310.1
8
1677
GenBank mRNA
AF105253.1
-
9
1621
AF493897.1
-
8
610
AF493898.1
-
8
568
AK026564.1
-
9
804
AK054862.1
-
6
1898
AK093534.1
-
7
844
AK122771.1
-
6
1017
AK225818.1
-
8
733
AK315860.1
-
8
532
AK315874.1
-
8
713
AL537118.3
-
8
730
BC002722.2
-
9
728
BC008855.2
-
10
755
BC022875.1
-
7
456
BC066923.1
-
7
568
BC089157.1
-
8
884
BC104928.1
-
8
656
BC108315.1
-
7
830
BT009905.1
-
8
610
KU178095.1
-
8
610
KU178096.1
-
8
568
M14631.1
-
9
580
X04408.1
-
8
678
X04409.1
-
8
580
X07036.1
-
8
622
X56009.1
-
8
686
Back To Top
More Information
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
endochondral ossification
energy reserve metabolic process
DNA methylation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
sensory perception of chemical stimulus
embryonic hindlimb morphogenesis
multicellular organism growth
post-embryonic body morphogenesis
response to drug
positive regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
developmental growth
embryonic cranial skeleton morphogenesis
cognition
cartilage development
bone development
hair follicle placode formation
platelet aggregation
genetic imprinting
regulation of parathyroid hormone secretion
GTPase activity
signal transducer activity
insulin-like growth factor receptor binding
protein binding
GTP binding
G-protein beta/gamma-subunit complex binding
beta-2 adrenergic receptor binding
D1 dopamine receptor binding
mu-type opioid receptor binding
ionotropic glutamate receptor binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0