Search
Search

GNB2| Gene Symbol: GNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005273.3 | NP_005264.2 | 8, 9 | 963 | ||
| GenBank mRNA | AF501883.1 | - | 7, 8 | 690 | ||
| AK056750.1 | - | 3, 4 | 1085 | |||
| AK291592.1 | - | 8, 9 | 838 | |||
| AL535249.3 | - | 8, 9 | 958 | |||
| BC010073.2 | - | 8, 9 | 887 | 4300639 | ||
| BC012348.1 | - | 8, 9 | 948 | 4556119 | ||
| BC068003.1 | - | 8, 9 | 873 | 6055863 | ||
| CR541729.1 | - | 7, 8 | 690 | |||
| M16538.1 | - | 8, 9 | 837 | |||
| M36429.1 | - | 7, 8 | 747 | |||