Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161415.1 | NP_001154887.1 | 4 | 1412 | ||
NM_001161416.1 | NP_001154888.1 | 3 | 1347 | |||
NM_001161417.1 | NP_001154889.1 | 2 | 1118 | |||
NM_005291.2 | NP_005282.1 | 3 | 1183 | |||
XM_017003833.1 | XP_016859322.1 | 2 | 1539 | |||
GenBank mRNA | AK126849.1 | - | 1 | 2373 | ||
AK292725.1 | - | 2 | 988 | |||
AK313949.1 | - | 2 | 874 | |||
BC031653.1 | - | 2 | 797 | 5174146 | ||
BC039595.1 | - | 2 | 802 | |||
JF810891.1 | - | 1 | 717 | |||
Y12546.1 | - | 3 | 875 | |||
Z94154.1 | - | 2 | 811 | |||
Z94155.1 | - | 1 | 595 |