Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSK3B SIRNA Assays ›
          Gene Symbol:
          GSK3B
          Gene Name:
          glycogen synthase kinase 3 beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 119821321 - 120094417 on Build GRCh38
          UniGene ID:
          Hs.445733
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS104523
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GSK3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146156.1 NP_001139628.1 5 1514
          NM_002093.3 NP_002084.2 5 1514
          XM_006713610.2 XP_006713673.1 5 1000
          XM_006713611.2 XP_006713674.1 5 1000
          GenBank mRNA AB451356.1 - 5 531
          AK290897.1 - 5 887
          BC000251.1 - 5 763 3357620
          BC012760.2 - 5 1513 3637163
          CR536510.1 - 5 531
          EU302497.1 - 5 531
          L33801.1 - 5 570

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          re-entry into mitotic cell cycle
          epithelial to mesenchymal transition
          positive regulation of cell-matrix adhesion
          glycogen metabolic process
          regulation of gene expression by genetic imprinting
          protein phosphorylation
          protein export from nucleus
          ER overload response
          dopamine receptor signaling pathway
          axonogenesis
          myoblast fusion
          aging
          circadian rhythm
          organ morphogenesis
          negative regulation of cardiac muscle hypertrophy
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of mitochondrial membrane potential
          negative regulation of neuron maturation
          response to activity
          Wnt signaling pathway
          cell migration
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          hippocampus development
          establishment of cell polarity
          negative regulation of protein complex assembly
          positive regulation of protein complex assembly
          negative regulation of protein binding
          positive regulation of protein binding
          response to estradiol
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          response to insulin
          regulation of microtubule-based process
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of smooth muscle cell apoptotic process
          protein localization to microtubule
          intracellular signal transduction
          cellular response to hepatocyte growth factor stimulus
          cellular response to interleukin-3
          cellular response to erythropoietin
          response to drug
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of MAP kinase activity
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          hypermethylation of CpG island
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          fat cell differentiation
          negative regulation of glycogen biosynthetic process
          positive regulation of protein catabolic process
          positive regulation of axon extension
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          positive regulation of protein export from nucleus
          regulation of neuronal synaptic plasticity
          positive regulation of smooth muscle cell proliferation
          negative regulation of dendrite morphogenesis
          negative regulation of nitric-oxide synthase activity
          negative regulation of NFAT protein import into nucleus
          canonical Wnt signaling pathway
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to hydrogen peroxide
          superior temporal gyrus development
          cellular response to mechanical stimulus
          cellular response to iron(II) ion
          cellular response to lithium ion
          response to epinephrine
          negative regulation of canonical Wnt signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          chemical synaptic transmission, postsynaptic
          regulation of cellular response to heat
          negative regulation of protein localization to nucleus
          positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          positive regulation of neuron death
          response to L-glutamate
          cellular response to dopamine
          negative regulation of dopaminergic neuron differentiation
          beta-catenin destruction complex assembly
          beta-catenin destruction complex disassembly
          response to insulin-like growth factor stimulus
          response to ultrasound
          response to angiotensin
          negative regulation of type B pancreatic cell development
          positive regulation of excitatory postsynaptic potential
          negative regulation of glycogen (starch) synthase activity
          positive regulation of DNA biosynthetic process
          positive regulation of stem cell differentiation
          negative regulation of neuron migration
          RNA polymerase II transcription factor binding
          p53 binding
          protein kinase activity
          protein serine/threonine kinase activity
          integrin binding
          protein binding
          ATP binding
          beta-catenin binding
          kinase activity
          protein kinase binding
          ubiquitin protein ligase binding
          protein kinase A catalytic subunit binding
          ionotropic glutamate receptor binding
          tau protein binding
          tau-protein kinase activity
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0