This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
GUCY2D
- Gene Name:
-
guanylate cyclase 2D, retinal
- Gene Aliases:
-
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC
- Chromosome Location:
-
Chr.17: 8002670 - 8020340 on Build GRCh38
- UniGene ID:
-
Hs.592109
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System