Search
Search

HAS1| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297436.1 | NP_001284365.1 | 4 | 1095 | ||
| NM_001523.3 | NP_001514.2 | 4 | 1098 | |||
| GenBank mRNA | AY916556.1 | - | 3 | 395 | ||
| BC035837.1 | - | 4 | 1077 | 5589083 | ||
| BX371636.2 | - | 2 | 190 | |||
| D84424.1 | - | 3 | 1082 | |||
| U59269.1 | - | 4 | 1074 | |||