Search
Search

HK1| Gene Symbol: HK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000188.2 | NP_000179.2 | 16 | 2362 | ||
| NM_001322364.1 | NP_001309293.1 | 18 | 2455 | |||
| NM_001322365.1 | NP_001309294.1 | 21 | 2760 | |||
| NM_001322366.1 | NP_001309295.1 | 16 | 2253 | |||
| NM_001322367.1 | NP_001309296.1 | 15 | 2266 | |||
| NM_033496.2 | NP_277031.1 | 16 | 2359 | |||
| NM_033497.2 | NP_277032.1 | 19 | 2577 | |||
| NM_033498.2 | NP_277033.1 | 19 | 2631 | |||
| NM_033500.2 | NP_277035.2 | 20 | 2724 | |||
| XM_005269737.1 | XP_005269794.1 | 16 | 2633 | |||
| XM_011539732.1 | XP_011538034.1 | 20 | 2631 | |||
| GenBank mRNA | AB209526.1 | - | 16 | 2358 | ||
| AK128226.1 | - | 21 | 2755 | |||
| AK292012.1 | - | 16 | 2362 | |||
| AK294428.1 | - | 16 | 2359 | |||
| BC008730.2 | - | 16 | 2317 | 3163058 | ||
| M75126.1 | - | 16 | 2342 | |||
| X66957.1 | - | 10 | 1403 | |||