Search
Search

ZBTB48| Gene Symbol: ZBTB48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278647.1 | NP_001265576.1 | 10 | 1876 | ||
| NM_001278648.1 | NP_001265577.1 | 10 | 1897 | |||
| NM_005341.3 | NP_005332.1 | 10 | 1963 | |||
| XM_017001112.1 | XP_016856601.1 | 10 | 2053 | |||
| XM_017001113.1 | XP_016856602.1 | 10 | 1994 | |||
| XM_017001114.1 | XP_016856603.1 | 9 | 1769 | |||
| GenBank mRNA | AF318381.1 | - | 3 | 2005 | ||
| AK290156.1 | - | 10 | 1871 | |||
| AK312786.1 | - | 10 | 1879 | |||
| BC013573.2 | - | 9 | 1822 | 3906038 | ||
| BX648395.1 | - | 6 | 2098 | |||
| L16896.1 | - | 10 | 1903 | |||