Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HOXA11 SIRNA Assays ›
Gene Symbol:
HOXA11
Gene Name:
homeobox A11
Gene Aliases:
HOX1, HOX1I, RUSAT1
Chromosome Location:
Chr.7: 27181157 - 27185216 on Build GRCh38
UniGene ID:
Hs.249171
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS104936
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HOXA11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005523.5
NP_005514.1
1
402
GenBank mRNA
AK313921.1
-
1
392
AL551705.3
-
1
415
BC040948.1
-
1
402
5587615
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109234, NM_001129878, XM_008762951, XM_003749752, BC166717.
This siRNA also targets the following transcripts in Mouse: NM_010450, AK133481, U20370, U20367, U20366, and others.
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
metanephros development
branching involved in ureteric bud morphogenesis
organ induction
transcription, DNA-templated
multicellular organism development
spermatogenesis
single fertilization
mesodermal cell fate specification
male gonad development
anatomical structure morphogenesis
anterior/posterior pattern specification
dorsal/ventral pattern formation
proximal/distal pattern formation
positive regulation of cell development
embryonic limb morphogenesis
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
embryonic digit morphogenesis
positive regulation of transcription, DNA-templated
developmental growth
uterus development
embryonic skeletal joint morphogenesis
cartilage development involved in endochondral bone morphogenesis
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cck9g:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0