Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000445.1 | - | 1 | 83 | ||
AK315194.1 | - | 1 | 100 | |||
BC032769.1 | - | 1 | 74 | |||
BC053894.1 | - | 1 | 135 | 6144625 |