Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000524.3 | NP_000515.2 | 1 | 1718 | ||
GenBank mRNA | AF498978.1 | - | 1 | 1145 | ||
AK291429.1 | - | 1 | 1379 | |||
AK314187.1 | - | 1 | 1559 | |||
BC069159.1 | - | 1 | 1145 | 7217020 | ||
BC136263.1 | - | 1 | 1235 | 9020250 | ||
DA160783.1 | - | 1 | 264 | |||
DA527925.1 | - | 1 | 20 |
Set Membership: |
G-protein coupled receptor |