Search
Search

IARS| Gene Symbol: IARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002161.5 | NP_002152.2 | 7 | 837 | ||
| NM_013417.3 | NP_038203.2 | 7 | 1007 | |||
| NR_073446.1 | 6 | 680 | ||||
| GenBank mRNA | AB209234.1 | - | 7 | 746 | ||
| AK293014.1 | - | 7 | 764 | |||
| AK301272.1 | - | 4 | 421 | |||
| BC065552.1 | - | 6 | 679 | 6043344 | ||
| BX537417.1 | - | 7 | 775 | |||
| BX537429.1 | - | 7 | 777 | |||
| BX538109.1 | - | 6 | 620 | |||
| BX647071.1 | - | 7 | 777 | |||
| BX647236.1 | - | 7 | 769 | |||
| BX647430.1 | - | 7 | 771 | |||
| BX648567.1 | - | 7 | 777 | |||
| D28473.1 | - | 7 | 925 | |||
| U04953.1 | - | 7 | 939 | |||