Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ID2 SIRNA Assays ›
          Gene Symbol:
          ID2 ID2B
          Gene Name:
          inhibitor of DNA binding 2, HLH protein
          inhibitor of DNA binding 2B, HLH protein (pseudogene)
          Gene Aliases:
          GIG8, ID2A, ID2H, bHLHb26
          -
          Chromosome Location:
          Chr.2: 8681983 - 8684453 on Build GRCh38
          Chromosome Location:
          Chr.3: 62123423 - 62124718 on Build GRCh38
          UniGene ID:
          Hs.180919
          Hs.591670
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS105176
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ID2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002166.4 NP_002157.2 1 393
          GenBank mRNA AK222682.1 - 1 320
          AK311988.1 - 1 320
          AY634687.1 - 1 329
          BC030639.2 - 1 320 4820416
          BE222494.1 - 1 393
          D13891.1 - 1 306
          Gene Symbol: ID2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M96843.1 - 1 319

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          metanephros development
          natural killer cell differentiation
          thigmotaxis
          membranous septum morphogenesis
          bundle of His development
          transcription, DNA-templated
          multicellular organism development
          adult locomotory behavior
          positive regulation of gene expression
          negative regulation of gene expression
          oligodendrocyte development
          regulation of lipid metabolic process
          olfactory bulb development
          circadian regulation of gene expression
          mammary gland epithelial cell proliferation
          regulation of circadian rhythm
          entrainment of circadian clock by photoperiod
          enucleate erythrocyte differentiation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          locomotor rhythm
          negative regulation of B cell differentiation
          positive regulation of fat cell differentiation
          positive regulation of erythrocyte differentiation
          positive regulation of macrophage differentiation
          negative regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of blood pressure
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          cell maturation
          Peyer's patch development
          embryonic digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          neuron fate commitment
          cell morphogenesis involved in neuron differentiation
          positive regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          adipose tissue development
          mammary gland alveolus development
          epithelial cell differentiation involved in mammary gland alveolus development
          endodermal digestive tract morphogenesis
          positive regulation of cell cycle arrest
          cellular response to lithium ion
          positive regulation of transcription involved in G1/S transition of mitotic cell cycle
          cellular senescence
          regulation of G1/S transition of mitotic cell cycle
          negative regulation of neural precursor cell proliferation
          protein binding
          ion channel binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0