Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000634.2 | NP_000625.1 | 2 | 1138 | ||
GenBank mRNA | AK298647.1 | - | 3 | 979 | ||
AK312668.1 | - | 2 | 1107 | |||
BC028221.1 | - | 2 | 1097 | 5217529 | ||
BC072397.1 | - | 2 | 1087 | 5755928 | ||
CR541994.1 | - | 1 | 1019 | |||
CR542029.1 | - | 1 | 1019 | |||
L19591.1 | - | 2 | 1138 | |||
M68932.1 | - | 2 | 1069 |