This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
ILF2
- Gene Name:
-
interleukin enhancer binding factor 2
- Gene Aliases:
-
NF45, PRO3063
- Chromosome Location:
-
Chr.1: 153661788 - 153671028 on Build GRCh38
- UniGene ID:
-
Hs.75117
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
ILF2 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001267809.1 |
NP_001254738.1 |
2, 3
|
181
|
|
| NM_004515.3 |
NP_004506.2 |
2, 3
|
181
|
|
| GenBank mRNA |
AK223326.1 |
- |
2, 3
|
101
|
|
| AK302214.1 |
- |
2, 3
|
125
|
|
| AK310615.1 |
- |
2, 3
|
145
|
|
| AK313364.1 |
- |
2, 3
|
140
|
|
| BC000382.2 |
- |
2, 3
|
106
|
2820505 |
| BM800544.1 |
- |
2, 3
|
156
|
|
| DC412988.1 |
- |
2, 3
|
181
|
|
| HQ322261.1 |
- |
1, 2
|
75
|
|
| HQ322262.1 |
- |
1, 2
|
75
|
|
| U10323.1 |
- |
2, 3
|
114
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System