Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002199.3 | NP_002190.2 | 6 | 647 | ||
GenBank mRNA | AK312953.1 | - | 6 | 625 | ||
BC015803.2 | - | 5 | 454 | 3920890 | ||
BT007264.1 | - | 5 | 447 | |||
CR457077.1 | - | 5 | 447 | |||
X15949.1 | - | 6 | 545 |