Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAK1 SIRNA Assays ›
          Gene Symbol:
          JAK1
          Gene Name:
          Janus kinase 1
          Gene Aliases:
          JAK1A, JAK1B, JTK3
          Chromosome Location:
          Chr.1: 64833223 - 65067746 on Build GRCh38
          UniGene ID:
          Hs.207538
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105629
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JAK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320923.1 NP_001307852.1 26 3629
          NM_001321852.1 NP_001308781.1 25 3632
          NM_001321853.1 NP_001308782.1 27 3891
          NM_001321854.1 NP_001308783.1 26 3807
          NM_001321855.1 NP_001308784.1 26 3810
          NM_001321856.1 NP_001308785.1 25 3545
          NM_001321857.1 NP_001308786.1 25 3723
          NM_002227.3 NP_002218.2 25 3726
          XM_017001212.1 XP_016856701.1 25 3570
          GenBank mRNA AB209057.1 - 24 3482
          AB219242.1 - 23 3373
          AK314257.1 - 26 3630
          BC132729.1 - 24 3444 40146751
          BX647675.1 - 11 1330
          HQ628631.1 - 23 3406
          M64174.1 - 24 3451

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell migration
          cell differentiation
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          interleukin-2-mediated signaling pathway
          regulation of cell proliferation
          positive regulation of GTPase activity
          innate immune response
          response to antibiotic
          interferon-gamma-mediated signaling pathway
          regulation of interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          positive regulation of sprouting angiogenesis
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          growth hormone receptor binding
          protein binding
          ATP binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          CCR5 chemokine receptor binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0