Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270422.1 | NP_001257351.1 | 6 | 1068 | ||
NM_001291622.1 | NP_001278551.1 | 6 | 1022 | |||
NM_001291623.1 | NP_001278552.1 | 3 | 815 | |||
NM_001291624.1 | NP_001278553.1 | 4 | 943 | |||
NM_001291625.1 | NP_001278554.1 | 5 | 1024 | |||
NM_018658.2 | NP_061128.2 | 5 | 940 | |||
NM_170741.2 | NP_733937.2 | 4 | 859 | |||
NM_170742.2 | NP_733938.2 | 4 | 896 | |||
XM_005257337.4 | XP_005257394.1 | 3 | 742 | |||
XM_006721885.3 | XP_006721948.1 | 7 | 1148 | |||
XM_006721886.3 | XP_006721949.1 | 6 | 1020 | |||
XM_006721887.3 | XP_006721950.1 | 6 | 1013 | |||
XM_011524781.2 | XP_011523083.1 | 5 | 979 | |||
XM_017024609.1 | XP_016880098.1 | 7 | 1251 | |||
XM_017024610.1 | XP_016880099.1 | 6 | 986 | |||
XM_017024611.1 | XP_016880100.1 | 6 | 1060 | |||
XM_017024612.1 | XP_016880101.1 | 5 | 979 | |||
XM_017024613.1 | XP_016880102.1 | 6 | 1107 | |||
GenBank mRNA | AF153815.1 | - | 5 | 880 | ||
AF153816.1 | - | 5 | 880 | |||
AF153817.1 | - | 3 | 652 | |||
AF179353.1 | - | 1 | 394 | |||
AK225944.1 | - | 8 | 905 | |||
AK290799.1 | - | 4 | 749 | |||
BC033038.2 | - | 4 | 886 | 5262150 |