Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIFC3 SIRNA Assays ›
          Gene Symbol:
          KIFC3
          Gene Name:
          kinesin family member C3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 57758217 - 57863053 on Build GRCh38
          UniGene ID:
          Hs.23131
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105813
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIFC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130099.1 NP_001123571.1 16 2396
          NM_001130100.1 NP_001123572.1 17 2452
          NM_001318710.1 NP_001305639.1 17 2333
          NM_001318711.1 NP_001305640.1 17 2494
          NM_001318712.1 NP_001305641.1 15 2069
          NM_001318713.1 NP_001305642.1 15 2462
          NM_001318714.1 NP_001305643.1 18 2841
          NM_001318715.1 NP_001305644.1 17 2430
          NM_005550.3 NP_005541.3 17 2452
          NR_134678.1 17 2734
          XM_005255937.1 XP_005255994.1 19 2961
          XM_005255944.1 XP_005256001.1 17 2315
          XM_006721188.1 XP_006721251.1 20 3039
          XM_011523075.1 XP_011521377.1 17 2619
          XM_011523076.1 XP_011521378.1 17 2619
          XM_011523077.1 XP_011521379.1 17 2418
          XM_011523078.1 XP_011521380.1 17 2221
          XM_011523079.1 XP_011521381.1 17 2556
          XM_017023219.1 XP_016878708.1 20 3008
          XM_017023220.1 XP_016878709.1 20 3185
          XM_017023221.1 XP_016878710.1 19 2961
          XM_017023222.1 XP_016878711.1 21 3203
          XM_017023223.1 XP_016878712.1 17 2336
          XM_017023224.1 XP_016878713.1 19 2870
          XM_017023225.1 XP_016878714.1 15 3440
          XM_017023226.1 XP_016878715.1 18 2713
          XM_017023227.1 XP_016878716.1 18 2713
          GenBank mRNA AB209290.1 - 17 2387
          AF004426.1 - 13 1838
          AK091891.1 - 2218
          AK291737.1 - 16 2291
          AK296995.1 - 17 2332
          AK299431.1 - 17 2394
          AK302743.1 - 15 2069
          AK303021.1 - 15 2390
          AK304132.1 - 17 2440
          AK307846.1 - 17 2410
          BC001211.1 - 17 2421 3503847
          BC008014.1 - 11 1417
          BC019057.2 - 4 421
          BC034234.1 - 18 2781 5296397
          BC041132.1 - 17 2282 5760988
          BC047051.1 - 17 2678 5297265
          BX647698.1 - 2 57

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          Golgi organization
          visual perception
          zonula adherens maintenance
          epithelial cell-cell adhesion
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, minus-end-directed
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0