Search
Search

KPNA5| Gene Symbol: KPNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002269.2 | NP_002260.2 | 3 | 290 | ||
| XM_011535805.1 | XP_011534107.1 | 4 | 468 | |||
| XM_017010838.1 | XP_016866327.1 | 4 | 468 | |||
| XM_017010839.1 | XP_016866328.1 | 3 | 271 | |||
| XM_017010840.1 | XP_016866329.1 | 4 | 428 | |||
| XM_017010842.1 | XP_016866331.1 | 3 | 271 | |||
| XM_017010843.1 | XP_016866332.1 | 4 | 395 | |||
| GenBank mRNA | AF005361.1 | - | 2 | 155 | ||
| AK314206.1 | - | 3 | 283 | |||
| BC047409.1 | - | 3 | 282 | 5297277 | ||
| BG721781.1 | - | 4 | 416 | |||