Search
Search

KRT84| Gene Symbol: KRT84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033045.3 | NP_149034.2 | 5 | 991 | ||
| XM_011538335.2 | XP_011536637.1 | 6 | 1106 | |||
| GenBank mRNA | AJ628420.1 | - | 5 | 937 | ||
| AK314030.1 | - | 5 | 1010 | |||
| BC069647.1 | - | 5 | 991 | 7262449 | ||